Assay Details
| Gene Symbol: CACNG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077832.1 | NP_001071300.1 | 2 - 3 | 399 | 81 | |
| GenBank mRNA | AF273330.1 | - | 2 - 3 | 303 | 81 | |
| BC126679.1 | - | 2 - 3 | 399 | 81 | ||
More Information
Set Membership: |
|

CACNG2
Genomic Map