Assay Details
| Gene Symbol: APOBEC3Z2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346123.1 | NP_001333052.1 | 2 - 3 | 309 | 88 | |
| GenBank mRNA | DQ974646.1 | - | 2 - 3 | 265 | 88 | |
| EU864535.1 | - | 2 - 3 | 312 | 88 | ||
| LC131309.1 | - | 2 - 3 | 211 | 88 | ||
More Information
Set Membership: |
|

APOBEC3Z2
Genomic Map