Assay Details
| Gene Symbol: APOBEC3Z2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ974646.1 | - | 4 - 5 | 655 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

APOBEC3Z2| Gene Symbol: APOBEC3Z2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ974646.1 | - | 4 - 5 | 655 | 72 | |
Set Membership: |
|