Assay Details
| Gene Symbol: APOBEC3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346124.1 | NP_001333053.1 | 2 - 3 | 309 | 88 | |
| GenBank mRNA | BC114797.1 | - | 2 - 3 | 293 | 88 | |
| EU864536.1 | - | 2 - 3 | 309 | 88 | ||
| LC131319.1 | - | 2 - 3 | 228 | 88 | ||
| Gene Symbol: APOBEC3Z2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ974646.1 | - | 5 - 6 | 819 | 88 | |
More Information
Set Membership: |
|

APOBEC3H
Genomic Map