Assay Details
| Gene Symbol: ABO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077926.1 | NP_001071394.1 | 7 - 8 | 478 | 67 | |
| GenBank mRNA | BC126634.1 | - | 7 - 8 | 478 | 67 | |
More Information
Set Membership: |
|

ABO
Genomic Map