Assay Details
| Gene Symbol: PELI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078004.1 | NP_001071472.1 | 2 - 3 | 466 | 81 | |
| GenBank mRNA | BC126542.1 | - | 2 - 3 | 466 | 81 | |
More Information
Set Membership: |
|

PELI1
Genomic Map