Assay Details
Gene Symbol: SCFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078005.1 | NP_001071473.1 | 6 - 7 | 1795 | 93 | |
GenBank mRNA | BC126501.1 | - | 6 - 7 | 1795 | 93 | |
BC149466.1 | - | 3 - 4 | 989 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
