Assay Details
| Gene Symbol: ELOVL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078042.1 | NP_001071510.1 | 2 - 3 | 146 | 66 | |
| GenBank mRNA | BC126611.1 | - | 2 - 3 | 146 | 66 | |
More Information
Set Membership: |
|

ELOVL7
Genomic Map