Assay Details
| Gene Symbol: ENC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078067.1 | NP_001071535.1 | 1 - 2 | 44 | 75 | |
| GenBank mRNA | BC126494.1 | - | 1 - 2 | 44 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC126494.1, NM_001078067.1 transcripts.
Set Membership: |
|

ENC1
Genomic Map