Assay Details
Gene Symbol: GADD45GIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080224.2 | NP_001073693.1 | 1 - 2 | 375 | 58 | |
GenBank mRNA | BC126785.1 | - | 1 - 2 | 376 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
