Assay Details
| Gene Symbol: MEPCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080283.2 | NP_001073752.2 | 3 - 4 | 2103 | 62 | |
| GenBank mRNA | BT029808.1 | - | 3 - 4 | 1332 | 62 | |
| DN530510.1 | - | 1 - 2 | 81 | 62 | ||
| EE245239.1 | - | 3 - 4 | 715 | 62 | ||
More Information
Set Membership: |
|

MEPCE
Genomic Map