Assay Details
| Gene Symbol: PRORSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080289.2 | NP_001073758.1 | 1 - 2 | 114 | 88 | |
| GenBank mRNA | BC126803.1 | - | 1 - 2 | 116 | 88 | |
More Information
Set Membership: |
|

PRORSD1
Genomic Map