Assay Details
| Gene Symbol: PCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080328.2 | NP_001073797.1 | 1 - 2 | 36 | 82 | |
| GenBank mRNA | BC126784.1 | - | 1 - 2 | 37 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC126784.1, NM_001080328.2 transcripts.
Set Membership: |
|

PCP2
Genomic Map