Assay Details
| Gene Symbol: PCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080328.2 | NP_001073797.1 | 3 - 4 | 302 | 63 | |
| GenBank mRNA | BC126784.1 | - | 3 - 4 | 303 | 63 | |
More Information
Set Membership: |
|

PCP2
Genomic Map