Assay Details
| Gene Symbol: THEM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080368.1 | NP_001073837.1 | 1 - 2 | 118 | 75 | |
| GenBank mRNA | BC126654.1 | - | 1 - 2 | 118 | 75 | |
More Information
Set Membership: |
|

THEM4
Genomic Map