Assay Details
Gene Symbol: TLCD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081516.1 | NP_001074985.1 | 3 - 4 | 556 | 70 | |
GenBank mRNA | BC133377.1 | - | 3 - 4 | 556 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
