Assay Details
| Gene Symbol: TMEM184B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081522.1 | NP_001074991.1 | 4 - 5 | 582 | 71 | |
| GenBank mRNA | BC133301.1 | - | 4 - 5 | 582 | 71 | |
More Information
Set Membership: |
|

TMEM184B
Genomic Map