Assay Details
Gene Symbol: PCMTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081526.1 | NP_001074995.1 | 6 - 7 | 1001 | 59 | |
GenBank mRNA | BC133335.1 | - | 6 - 7 | 1001 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
