Assay Details
| Gene Symbol: C16H1orf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081547.1 | NP_001075016.1 | 1 - 2 | 277 | 86 | |
| GenBank mRNA | BC133418.1 | - | 1 - 2 | 277 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC133418.1, NM_001081547.1 transcripts.
Set Membership: |
|

C16H1orf21
Genomic Map