Assay Details
| Gene Symbol: C16H1orf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081547.1 | NP_001075016.1 | 2 - 3 | 503 | 70 | |
| GenBank mRNA | BC133418.1 | - | 2 - 3 | 503 | 70 | |
More Information
Set Membership: |
|

C16H1orf21
Genomic Map