Assay Details
Gene Symbol: DES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081575.1 | NP_001075044.1 | 5 - 6 | 1089 | 65 | |
GenBank mRNA | AB011673.1 | - | 5 - 6 | 998 | 65 | |
BC133410.1 | - | 5 - 6 | 1089 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
