Assay Details
| Gene Symbol: HPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081593.1 | NP_001075062.1 | 7 - 8 | 984 | 133 | |
| GenBank mRNA | BC133472.1 | - | 7 - 8 | 984 | 133 | |
More Information
Set Membership: |
|

HPF1
Genomic Map