Assay Details
| Gene Symbol: ELK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081612.1 | NP_001075081.1 | 4 - 5 | 1387 | 65 | |
| GenBank mRNA | BC133356.1 | - | 4 - 5 | 1387 | 65 | |
More Information
Set Membership: |
|

ELK4
Genomic Map