Assay Details
Gene Symbol: TCF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081721.2 | NP_001075190.1 | 1 - 2 | 609 | 115 | |
GenBank mRNA | BC133638.1 | - | 1 - 2 | 610 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
