Assay Details
Gene Symbol: DDX49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081729.1 | NP_001075198.1 | 6 - 7 | 732 | 54 | |
GenBank mRNA | BT029794.1 | - | 6 - 7 | 732 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
