Assay Details
| Gene Symbol: PRSS45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082455.2 | NP_001075924.1 | 5 - 6 | 776 | 137 | |
| GenBank mRNA | BC133553.1 | - | 5 - 6 | 777 | 137 | |
More Information
Set Membership: |
|

PRSS45
Genomic Map