Assay Details
| Gene Symbol: RPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082605.2 | NP_001076074.1 | 8 - 9 | 1416 | 65 | |
| GenBank mRNA | BC133459.1 | - | 8 - 9 | 1417 | 65 | |
More Information
Set Membership: |
|

RPN1
Genomic Map