Assay Details
Gene Symbol: CCNI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083373.1 | NP_001076842.1 | 6 - 7 | 1269 | 71 | |
GenBank mRNA | BC114754.1 | - | 6 - 7 | 1269 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
