Assay Details
| Gene Symbol: NUDT17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083383.2 | NP_001076852.1 | 7 - 8 | 908 | 70 | |
| GenBank mRNA | BC114872.1 | - | 7 - 8 | 909 | 70 | |
More Information
Set Membership: |
|

NUDT17
Genomic Map