Assay Details
Gene Symbol: FAM83D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083393.1 | NP_001076862.1 | 1 - 2 | 510 | 136 | |
GenBank mRNA | BC133491.1 | - | 1 - 2 | 510 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
