Assay Details
| Gene Symbol: RBBP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083424.1 | NP_001076893.1 | 5 - 6 | 537 | 67 | |
| GenBank mRNA | BC123685.1 | - | 5 - 6 | 537 | 67 | |
More Information
Set Membership: |
|

RBBP9
Genomic Map