Assay Details
| Gene Symbol: BRMS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083428.1 | NP_001076897.1 | 7 - 8 | 709 | 136 | |
| GenBank mRNA | BC123661.1 | - | 7 - 8 | 709 | 136 | |
More Information
Set Membership: |
|

BRMS1L
Genomic Map