Assay Details
| Gene Symbol: SEC22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083462.1 | NP_001076931.1 | 3 - 4 | 557 | 85 | |
| GenBank mRNA | BC114842.1 | - | 4 - 5 | 584 | 85 | |
More Information
Set Membership: |
|

SEC22A
Genomic Map