Assay Details
Gene Symbol: CAPZA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083480.2 | NP_001076949.1 | 1 - 2 | 82 | 63 | |
GenBank mRNA | BC114648.1 | - | 1 - 2 | 86 | 63 | |
BT030727.1 | - | 1 - 2 | 43 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
