Assay Details
| Gene Symbol: RAMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083505.2 | NP_001076974.1 | 1 - 2 | 94 | 92 | |
| GenBank mRNA | BC114856.1 | - | 1 - 2 | 95 | 92 | |
More Information
Set Membership: |
|

RAMP3
Genomic Map