Assay Details
Gene Symbol: NEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083642.1 | NP_001077111.1 | 1 - 2 | 263 | 63 | |
GenBank mRNA | BC134571.1 | - | 1 - 2 | 263 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
