Assay Details
| Gene Symbol: RASL10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083645.1 | NP_001077114.1 | 1 - 2 | 630 | 102 | |
| GenBank mRNA | BC134570.1 | - | 1 - 2 | 630 | 102 | |
More Information
Set Membership: |
|

RASL10A
Genomic Map