Assay Details
| Gene Symbol: LOC513767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083681.2 | NP_001077150.1 | 5 - 6 | 687 | 79 | |
| GenBank mRNA | BC134787.1 | - | 5 - 6 | 688 | 79 | |
More Information
Set Membership: |
|

LOC513767
Genomic Map