Assay Details
Gene Symbol: FRRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083695.2 | NP_001077164.1 | 13 - 14 | 1589 | 63 | |
GenBank mRNA | BC133504.1 | - | 13 - 14 | 1592 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
