Assay Details
Gene Symbol: KLF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083727.1 | NP_001077196.1 | 3 - 4 | 1493 | 97 | |
GenBank mRNA | BC134493.1 | - | 3 - 4 | 1493 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
