Assay Details
Gene Symbol: GEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083732.2 | NP_001077201.2 | 3 - 4 | 604 | 63 | |
NM_001276305.1 | NP_001263234.1 | 3 - 4 | 530 | 63 | ||
GenBank mRNA | BC134475.1 | - | 3 - 4 | 605 | 63 | |
DV883786.1 | - | 3 - 4 | 476 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
