Assay Details
| Gene Symbol: JAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083736.1 | NP_001077205.1 | 4 - 5 | 653 | 66 | |
| GenBank mRNA | BC134600.1 | - | 4 - 5 | 653 | 66 | |
More Information
Set Membership: |
|

JAM2
Genomic Map