Assay Details
| Gene Symbol: GPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083740.2 | NP_001077209.1 | 10 - 11 | 1547 | 65 | |
| GenBank mRNA | BC134583.1 | - | 10 - 11 | 1548 | 65 | |
More Information
Set Membership: |
|

GPT
Genomic Map