Assay Details
| Gene Symbol: AUNIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083742.1 | NP_001077211.1 | 2 - 3 | 322 | 100 | |
| GenBank mRNA | BC134777.1 | - | 2 - 3 | 322 | 100 | |
More Information
Set Membership: |
|

AUNIP
Genomic Map