Assay Details
| Gene Symbol: GFRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083747.1 | NP_001077216.1 | 4 - 5 | 822 | 67 | |
| GenBank mRNA | BC134488.1 | - | 4 - 5 | 822 | 67 | |
More Information
Set Membership: |
|

GFRA3
Genomic Map