Assay Details
Gene Symbol: CMTM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083749.1 | NP_001077218.1 | 3 - 4 | 625 | 59 | |
GenBank mRNA | BC134504.1 | - | 3 - 4 | 625 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
