Assay Details
Gene Symbol: SYNPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083751.1 | NP_001077220.1 | 1 - 2 | 266 | 56 | |
GenBank mRNA | BC134621.1 | - | 1 - 2 | 266 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
