Assay Details
| Gene Symbol: LPGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083756.1 | NP_001077225.1 | 8 - 9 | 1234 | 83 | |
| GenBank mRNA | BC134696.1 | - | 8 - 9 | 1234 | 83 | |
More Information
Set Membership: |
|

LPGAT1
Genomic Map