Assay Details
| Gene Symbol: LY86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083761.1 | NP_001077230.1 | 5 - 6 | 554 | 83 | |
| GenBank mRNA | BC134752.1 | - | 5 - 6 | 554 | 83 | |
More Information
Set Membership: |
|

LY86
Genomic Map