Assay Details
Gene Symbol: LSM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083781.1 | NP_001077250.1 | 1 - 2 | 283 | 75 | |
GenBank mRNA | BC134669.1 | - | 1 - 2 | 283 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC134669.1, NM_001083781.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
